2,3-Dihydroxybenzoic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Pyrocatechuic acid
A10144.09 A10144.18 A10144.30
A10144.09EA 1850 SEK
A10144.09 A10144.18 A10144.30
2,3-Dihydroxybenzoic acid ≥98%
2,3-Dihydroxybenzoic acid
Purity: 98% CAS No.: 303-38-8 Molecular Formula: C7H6O4 Formula Weight: 154.12 Melting Point: 204-208° Beilstein Reference: 2209117
Formel: C₇H₆O₄
MW: 154,12 g/mol
Kokpunkt: 339 °C (1013 hPa)
Smältpunkt: 204…208 °C
Densitet: 1,542
Storage Temperature: Omgivande
MDL Number: MFCD00002446
CAS nummer: 303-38-8
EINECS: 206-139-5