3,4,5-Trimethoxybenzoic acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: Trimethylgallic Acid
139930050 139931000 139935000
ACRO139930050EA 395 SEK
ACRO139930050 ACRO139931000 ACRO139935000
3,4,5-Trimethoxybenzoic acid 99%
3,4,5-Trimethoxybenzoic acid
Formel: C₁₀H₁₂O₅
MW: 212,2 g/mol
Kokpunkt: 225…227 °C (10 mmHg)
Smältpunkt: 168…172 °C
Densitet: 1,31 g/cm³ (24.95 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002501
CAS nummer: 118-41-2
EINECS: 204-248-2
Order Now
Specifikation testresultat
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 172 °C
HPLC ≥98.5 %