3,4,5-Triethoxybenzoic acid ≥98%
Leverantör: Thermo Scientific
Varning
A14157.14 A14157.22 A14157.36
A14157.14EA 610 SEK
A14157.14 A14157.22 A14157.36
3,4,5-Triethoxybenzoic acid ≥98%
3,4,5-Triethoxybenzoic acid
Purity: 98+% CAS No.: 6970-19-0 Molecular Formula: C13H18O5 Formula Weight: 254.29 Melting Point: 110-113° Beilstein Reference: 2659063
Formel: C₁₃H₁₈O₅
MW: 254,28 g/mol
Kokpunkt: 375 °C (1013 hPa)
Smältpunkt: 110…113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002505
CAS nummer: 6970-19-0
EINECS: 230-192-3