p-Anisic acid 98
Leverantör: Thermo Fisher Scientific
Synonyms: 4-Methoxybenzoic acid, 4-Anisic acid
A15697.0B A15697.30
A15697.0BEA 2700 SEK
A15697.0B A15697.30
p-Anisic acid 98
p-Anisic acid
Purity: 98+% CAS No.: 100-09-4 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 185°(365°F) Odour: Odourless pH: 3-4 Melting Point: 182-185° Boiling Point: 275-277° Beilstein Reference: 508910 Merck Reference: 14,666 MDL No.: MFCD00002542
Formel: C₈H₈O₃
MW: 152.15 g/mol
Kokpunkt: 275…277 °C
Smältpunkt: 182…185 °C
Densitet: 1.385
Flampunkt: 185 °C (365 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002542
CAS nummer: 100-09-4
EINECS: 202-818-5
Merck Index: 13,00669