4-Dodecyloxybenzoic acid ≥98%

Leverantör: Thermo Scientific
Varning

Synonyms: 4-(Dodecyloxy)benzoic acid

B22689.06 B22689.14
B22689.06EA 865 SEK
B22689.06 B22689.14
4-Dodecyloxybenzoic acid ≥98%
4-Dodecyloxybenzoic acid

Purity: 98% CAS No.: 2312-15-4 Molecular Formula: C19H30O3 Formula Weight: 306.45 Melting Point: 132-137° Beilstein Reference: 2215914

Formel: C₁₉H₃₀O₃
MW: 306,45 g/mol
Kokpunkt: 420 °C (1013 hPa)
Smältpunkt: 132…137 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002543
CAS nummer: 2312-15-4
EINECS: 219-004-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR