4-Hydroxy-3-nitrobenzoic acid ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A10562.14 A10562.22
A10562.14EA 850 SEK
A10562.14 A10562.22
4-Hydroxy-3-nitrobenzoic acid ≥98%
4-Hydroxy-3-nitrobenzoic acid
Purity: 98% CAS No.: 616-82-0 Molecular Formula: C7H5NO5 Formula Weight: 183.12 Melting Point: 182-184° Beilstein Reference: 2213134
Formel: C₇H₅NO₅
MW: 183.12 g/mol
Smältpunkt: 182…184 °C
Densitet: 1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007119
CAS nummer: 616-82-0
EINECS: 210-494-1