5-Hydroxy-2-nitrobenzoic acid

Leverantör: Thermo Scientific
Varning

RH00465DA RH00465EA RH00465EE
MAYBRH00465DAEA 0 SEK
MAYBRH00465DA MAYBRH00465EA MAYBRH00465EE
5-Hydroxy-2-nitrobenzoic acid
5-Hydroxy-2-nitrobenzoic acid
Formel: C₇H₅NO₅
MW: 183,12 g/mol
MDL Number: MFCD00017566
CAS nummer: 610-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR