3-Nitrobenzoic acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: m-Nitrobenzoic acid
128455000 128450051 128451000
ACRO128455000EA 1100 SEK
ACRO128455000 ACRO128450051 ACRO128451000
3-Nitrobenzoic acid 99%
3-Nitrobenzoic acid
Formel: C₇H₅NO₄
MW: 167,12 g/mol
Kokpunkt: 341 °C (1013 hPa)
Smältpunkt: 139…143 °C
Densitet: 1,494 g/cm³ (20 °C)
Flampunkt: 190 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007251
CAS nummer: 121-92-6
EINECS: 204-508-5
Merck Index: 12,06686
Order Now
Specifikation testresultat
Appearance White to light yellow or light green Powder or crystals
Infrared spectrum Conforms
Melting point 139 to 143 °C
HPLC ≥98.5 %
Titration with NaOH 98.5 to 101.5 %
Specific optical rotation -56° to -61° (20 °C, 589 nm) (c=1, chloroform)