5-Aminosalicylsyra 95%
Leverantör: Thermo Scientific
Varning
Synonyms: Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid
B23970.22 B23970.36
B23970.22EA 1250 SEK
B23970.22 B23970.36
5-Aminosalicylsyra 95%
5-Aminosalicylsyra
Purity: 95% CAS No.: 89-57-6 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: ca 280° dec. Beilstein Reference: 2090421 Merck Reference: 14,5904 MDL No.: MFCD00007877
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Kokpunkt: 347 °C (1013 hPa)
Smältpunkt: ∼280 °C (dec.)
Densitet: 1,568 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00007877
CAS nummer: 89-57-6
EINECS: 201-919-1
Merck Index: 14,5904