5-Aminosalicylsyra 99%
Leverantör: Thermo Scientific
Varning
Synonyms: Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid
134330050 134330250 134331000
ACRO134330050EA 455 SEK
ACRO134330050 ACRO134330250 ACRO134331000
5-Aminosalicylsyra 99%
5-Aminosalicylsyra
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Kokpunkt: 347 °C (1013 hPa)
Smältpunkt: 283 °C (dec.)
Densitet: 1,568 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00007877
CAS nummer: 89-57-6
EINECS: 201-919-1
Merck Index: 12,05964
Order Now
Specifikation testresultat
Appearance Beige to grey to light brown Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 3 hrs)
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 % (On dry substance)