3-Amino-4-hydroxybenzoic acid 98%
Leverantör: Thermo Fisher Scientific
Varning
271490050 271490250
ACRO271490050EA 700 SEK
ACRO271490050 ACRO271490250
3-Amino-4-hydroxybenzoic acid 98%
3-Amino-4-hydroxybenzoic acid
Formel: C₇H₇NO₃
Kokpunkt: 347 °C (1013 hPa)
Smältpunkt: 198…205 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007697
CAS nummer: 1571-72-8
EINECS: 216-390-2
Order Now
Specifikation testresultat
Appearance Beige to grey to brown Crystalline powder
Infrared spectrum Conforms
Melting point 198 to 205 °C (Dec)
HPLC ≥97.5 %