2-Amino-5-hydroxybenzoic acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Hydroxyanthranilic acid
310310100 310310010
ACRO310310100EA 1225 SEK
ACRO310310100 ACRO310310010
2-Amino-5-hydroxybenzoic acid 99%
2-Amino-5-hydroxybenzoic acid
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Kokpunkt: 347 °C (1013 hPa)
Smältpunkt: 233…234 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007870
CAS nummer: 394-31-0
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Grey-brown Crystalline powder
Infrared spectrum Conforms
Titration with TBAH ≥98.5 %