4-aminobensoesyra 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Vitamin Bx, para-Aminobenzoic acid, PABA, Vitamin H1
A12673.0B A12673.18 A12673.30 A12673.PG
A12673.0BEA 3500 SEK
A12673.0B A12673.18 A12673.30 A12673.PG
4-aminobensoesyra 99%
4-aminobensoesyra
Purity: 99% CAS No.: 150-13-0 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Melting Point: ca 187° dec. Beilstein Reference: 471605 Merck Reference: 14,423 MDL No.: MFCD00007894
Formel: C₇H₇NO₂
Kokpunkt: 340 °C (1013 hPa)
Smältpunkt: ∼187 °C (dec.)
Densitet: 1,374
Storage Temperature: Omgivande
MDL Number: MFCD00007894
CAS nummer: 150-13-0
EINECS: 205-753-0
Merck Index: 14,423