Natriumbensoat, RonaCare®
Leverantör: Merck
|
Varning
|
Synonyms:
Benzoic acid sodium salt
1.30176.9026
1.30176.9026EA
0
SEK
1.30176.9026
Natriumbensoat, RonaCare®
Natriumbensoat
Formel:
C₆H₅COONa MW: 144,11 g/mol Smältpunkt: 410…430 °C Densitet: 1,44 g/cm³ (20 °C) Flampunkt: >100 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00012463 CAS nummer: 532-32-1 EINECS: 208-534-8 REACH: 01-2119460683-35 Merck Index: 13,08654 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...