4-Hydroxybenzonitrile ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A15803.18 A15803.22 A15803.30 A15803.36
A15803.18EA 1575 SEK
A15803.18 A15803.22 A15803.30 A15803.36
4-Hydroxybenzonitrile ≥98%
4-Hydroxybenzonitrile
Purity: 98+% CAS No.: 767-00-0 Molecular Formula: C7H5NO Formula Weight: 119.12 Melting Point: 110-114° Beilstein Reference: 386130
Formel: C₇H₅NO
Kokpunkt: 148 °C (1 mmHg)
Smältpunkt: 110…114 °C
Densitet: ∼1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002312
CAS nummer: 767-00-0
EINECS: 212-175-2