3-Hydroxybenzonitrile 97%
Leverantör: Thermo Scientific
Varning
Synonyms: m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol
110820100 110820500
ACRO110820100EA 1700 SEK
ACRO110820100 ACRO110820500
3-Hydroxybenzonitrile 97%
3-Hydroxybenzonitrile
Formel: C₇H₅NO
MW: 119,12 g/mol
Kokpunkt: 265 °C (1013 hPa)
Smältpunkt: 75…80 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002252
CAS nummer: 873-62-1
EINECS: 212-845-4
UN: 3439
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 75 to 80 °C
GC ≥96.0 %