3-Methoxybenzonitrile ≥98%
Leverantör: Thermo Scientific
Fara
B24171.06 B24171.14 B24171.22
B24171.06EA 785 SEK
B24171.06 B24171.14 B24171.22
3-Methoxybenzonitrile ≥98%
3-Methoxybenzonitrile
Purity: 98% CAS No.: 1527-89-5 Molecular Formula: C8H7NO Formula Weight: 133.15 Flash Point: 105°(221°F) Refractive Index: 1.5402 Odour: Almond-like Melting Point: 22-24° Boiling Point: 110-111°/12mm Beilstein Reference: 1932666 UN No.: 3439 MDL No.: MFCD00001801
Formel: C₈H₇NO
MW: 133,15 g/mol
Kokpunkt: 110…111 °C (12 mmHg)
Smältpunkt: 22…24 °C
Densitet: 1,089
Flampunkt: 105 °C (221 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001801
CAS nummer: 1527-89-5
EINECS: 216-201-3
UN: 3439
ADR: 6.1,III