2-Methoxybenzonitrile ≥98%
Leverantör: Thermo Scientific
|
Fara
|
Synonyms:
o-Cyanoanisol, 2-Cyanoanisol, o-Methoxybenzonitrile
L01293.06
L01293.14
L01293.22
L01293.06EA
965
SEK
L01293.06
L01293.14
L01293.22
2-Methoxybenzonitrile ≥98%
2-Methoxybenzonitrile
Purity: 98%
CAS No.: 6609-56-9
Molecular Formula: C8H7NO
Formula Weight: 133.15
Flash Point:
Formel:
C₈H₇NO MW: 133,15 g/mol Kokpunkt: 256 °C Smältpunkt: 24,5 °C Densitet: 1,093 Flampunkt: >110 °C (230 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00001783 CAS nummer: 6609-56-9 EINECS: 229-559-0 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...