Benzonitril 99+% Spectroscopy
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Cyanobenzene, Phenylcyanide
167672500 167670010 167670500
ACRO167672500EA 1325 SEK
ACRO167672500 ACRO167670010 ACRO167670500
Benzonitril 99+% Spectroscopy
Benzonitril
Formel: C₇H₅N
MW: 103.12 g/mol
Kokpunkt: 188…191 °C (760 mmHg)
Smältpunkt: –13 °C
Densitet: 1.01 g/cm³
Flampunkt: 71 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001770
CAS nummer: 100-47-0
EINECS: 202-855-7
UN: 2224
ADR: 6.1 II
Merck Index: 13,01098
Order Now
Specifikation testresultat
Appearance Clear liquid
Water ≤0.03 %
GC ≥99.0 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %