3-Hydroxybenzaldehyde 98.5%
Leverantör: Thermo Scientific
Varning
120961000 120965000
ACRO120961000EA 985 SEK
ACRO120961000 ACRO120965000
3-Hydroxybenzaldehyde 98.5%
3-Hydroxybenzaldehyde
Formel: C₇H₆O₂
MW: 122,12 g/mol
Kokpunkt: 191 °C (50 mmHg)
Smältpunkt: 101…105 °C
Densitet: 1,1299 g/cm³ (114 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003368
CAS nummer: 100-83-4
EINECS: 202-892-9
Order Now
Specifikation testresultat
Appearance Light yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 101 to 105 °C
GC ≥98.0 %