3,4,5-Trimethoxybenzaldehyde 99%
Leverantör: Thermo Scientific
Varning
139910250 139911000 139915000
ACRO139910250EA 535 SEK
ACRO139910250 ACRO139911000 ACRO139915000
3,4,5-Trimethoxybenzaldehyde 99%
3,4,5-Trimethoxybenzaldehyde
Formel: C₁₀H₁₂O₄
MW: 196,2 g/mol
Kokpunkt: 163…165 °C (10 mmHg)
Smältpunkt: 73…76 °C
Densitet: 1,367 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003364
CAS nummer: 86-81-7
EINECS: 201-701-6
UN: 2811
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to cream Powder or flakes
Infrared spectrum Conforms
Melting point 73 to 76 °C
GC ≥96.0 %