2,4,6-Trimethoxybenzaldehyde ≥98%
Leverantör: Thermo Scientific
Varning
A11238.09 A11238.18
A11238.09EA 1450 SEK
A11238.09 A11238.18
2,4,6-Trimethoxybenzaldehyde ≥98%
2,4,6-Trimethoxybenzaldehyde
Purity: 98% CAS No.: 830-79-5 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 119-123° Beilstein Reference: 1956051
Formel: C₁₀H₁₂O₄
MW: 196,2 g/mol
Kokpunkt: 291 °C (1013 hPa)
Smältpunkt: 119…123 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003313
CAS nummer: 830-79-5
EINECS: 212-598-2