Syringaldehyde ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,5-Dimethoxy-4-hydroxybenzaldehyde, 4-Hydroxy-3,5-dimethoxybenzaldehyde
A12933.06 A12933.14 A12933.22
A12933.06EA 670 SEK
A12933.06 A12933.14 A12933.22
Syringaldehyde ≥98%
Syringaldehyde
Purity: 98+% CAS No.: 134-96-3 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 110-113° Boiling Point: 192-193°/14mm Merck Reference: 14,9015 MDL No.: MFCD00006943
Formel: C₉H₁₀O₄
MW: 182,18 g/mol
Kokpunkt: 192…193 °C (14 mmHg)
Smältpunkt: 110…113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006943
CAS nummer: 134-96-3
EINECS: 205-167-5
Merck Index: 14,9015