2,4-Dimethoxybenzaldehyde ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A12549.14 A12549.22
A12549.14EA 870 SEK
A12549.14 A12549.22
2,4-Dimethoxybenzaldehyde ≥98%
2,4-Dimethoxybenzaldehyde
Purity: 98% CAS No.: 613-45-6 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 67-71° Boiling Point: 165°/10mm MDL No.: MFCD00003311
Formel: C₉H₁₀O₃
Kokpunkt: 165 °C (10 mmHg)
Smältpunkt: 67…71 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003311
CAS nummer: 613-45-6
EINECS: 210-342-4