2-Hydroxy-m-anisaldehyde 99
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: o-Vanillin, 2-Hydroxy-3-methoxybenzaldehyde
A15672.14 A15672.22 A15672.36
A15672.14EA 590 SEK
A15672.14 A15672.22 A15672.36
2-Hydroxy-m-anisaldehyde 99
2-Hydroxy-m-anisaldehyde
Purity: 99% CAS No.: 148-53-8 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 40-42° Boiling Point: 265-266° MDL No.: MFCD00003322
Formel: C₈H₈O₃
MW: 152.15 g/mol
Kokpunkt: 265…266 °C
Smältpunkt: 40…42 °C
Densitet: 1.2039 g/cm³ (45 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003322
CAS nummer: 148-53-8
EINECS: 205-715-3