2-Hydroxy-m-anisaldehyde 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: o-Vanillin, 2-Hydroxy-3-methoxybenzaldehyde
153071000 153075000 153070100
ACRO153071000EA 970 SEK
ACRO153071000 ACRO153075000 ACRO153070100
2-Hydroxy-m-anisaldehyde 99%
2-Hydroxy-m-anisaldehyde
Formel: C₈H₈O₃
Kokpunkt: 265…266 °C (760 mmHg)
Smältpunkt: 40…42 °C
Densitet: 1,2039 g/cm³ (45 °C)
Flampunkt: >112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003322
CAS nummer: 148-53-8
EINECS: 205-715-3
Order Now
Specifikation testresultat
Appearance Yellow to brown Crystals or flakes or crystalline mass
Infrared spectrum Conforms
Melting point 42 to 45 °C
GC ≥98.5 %
HPLC ≥98.5 %