4-Ethoxybenzaldehyde 99%
Leverantör: Thermo Fisher Scientific
Varning
174250250 174251000
ACRO174250250EA 620 SEK
ACRO174250250 ACRO174251000
4-Ethoxybenzaldehyde 99%
4-Ethoxybenzaldehyde
Formel: C₉H₁₀O₂
Kokpunkt: 255 °C (760 mmHg)
Smältpunkt: 13…14 °C
Densitet: 1,08 g/cm³
Flampunkt: 75 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003388
CAS nummer: 10031-82-0
EINECS: 233-093-3
Order Now
Specifikation testresultat
Appearance Clear yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5574 to 1.5594 (20°C, 589 nm)
GC ≥98.5 %