p-Dimetylaminobensaldehyd 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
A11712.0E A11712.22 A11712.36
A11712.0EEA 10000 SEK
A11712.0E A11712.22 A11712.36
p-Dimetylaminobensaldehyd 98%
p-Dimetylaminobensaldehyd
Purity: 98% CAS No.: 100-10-7 Molecular Formula: C9H11NO Formula Weight: 149.19 Flash Point: 164°(327°F) Odour: Strong Boiling Point: 176-177°/17mm Beilstein Reference: 606802 Merck Reference: 14,3230 MDL No.: MFCD00003381
Formel: (CH₃)₂NC₆H₄CHO
Kokpunkt: 176…177 °C (17 mmHg)
Smältpunkt: 73…75 °C
Densitet: 1,0254 g/cm³ (99.9 °C)
Flampunkt: 164 °C (327 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003381
CAS nummer: 100-10-7
EINECS: 202-819-0
Merck Index: 14,3230
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f59709e90756a237e9c4bd808e8fd50ac9367d91&vid=909765f6b48eb3673cc290753e0ead3358b739cf&prid=web&clid=SAPDAM