2,6-Dibromophenol ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,3-Dibromo-2-hydroxybenzene
A10573.03 A10573.06 A10573.14
A10573.03EA 875 SEK
A10573.03 A10573.06 A10573.14
2,6-Dibromophenol ≥99%
2,6-Dibromophenol
Purity: 99% CAS No.: 608-33-3 Molecular Formula: C6H4Br2O Formula Weight: 251.92 Odour: Phenol-like Melting Point: 53-57° Boiling Point: 255-256° Beilstein Reference: 2043614 MDL No.: MFCD00002152
Formel: C₆H₄Br₂O
MW: 251,91 g/mol
Kokpunkt: 255…256 °C
Smältpunkt: 53…57 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002152
CAS nummer: 608-33-3
EINECS: 210-161-0
UN: 2811
ADR: 6.1,III