2-Bromo-1,4-dimethoxybenzene ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Bromo-2,5-dimethoxybenzene
A15640.09 A15640.18 A15640.30
A15640.09EA 1175 SEK
A15640.09 A15640.18 A15640.30
2-Bromo-1,4-dimethoxybenzene ≥98%
2-Bromo-1,4-dimethoxybenzene
Purity: 98% CAS No.: 25245-34-5 Molecular Formula: C8H9BrO2 Formula Weight: 217.07 Flash Point: 110°(230°F) Refractive Index: 1.5700 Boiling Point: 260-262° Beilstein Reference: 2441884 MDL No.: MFCD00008355
Formel: C₈H₉BrO₂
Kokpunkt: 260…262 °C
Densitet: 1,441
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008355
CAS nummer: 25245-34-5
EINECS: 246-756-7