2-Bromoanisole ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Bromo-2-methoxybenzene
A11692.14 A11692.22 A11692.36
A11692.14EA 860 SEK
A11692.14 A11692.22 A11692.36
2-Bromoanisole ≥98%
2-Bromoanisole
Purity: 98% CAS No.: 578-57-4 Molecular Formula: C7H7BrO Formula Weight: 187.04 Flash Point: 96°(204°F) Refractive Index: 1.5740 Melting Point: 2-3° Boiling Point: 220-222° Beilstein Reference: 1859996 MDL No.: MFCD00000064
Formel: C₇H₇BrO
MW: 187,04 g/mol
Kokpunkt: 220…222 °C
Smältpunkt: 2…3 °C
Densitet: 1,511
Flampunkt: 96 °C (204 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000064
CAS nummer: 578-57-4
EINECS: 209-425-8
UN: 3082
ADR: 9,III