1-Bromo-3-nitrobenzene 99%
Leverantör: Thermo Scientific
Varning
160390250 160391000
ACRO160390250EA 1325 SEK
ACRO160390250 ACRO160391000
1-Bromo-3-nitrobenzene 99%
1-Bromo-3-nitrobenzene
Formel: C₆H₄BrNO₂
MW: 202,01 g/mol
Kokpunkt: 256 °C (1013 hPa)
Smältpunkt: 51…54 °C
Densitet: 1,704 g/cm³
Flampunkt: 113 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00024298
CAS nummer: 585-79-5
EINECS: 209-563-9
UN: 3459
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to yellow to brown Crystalline powder or crystals or solid
Infrared spectrum Conforms
Melting point 51 to 55 °C
GC ≥98.5 %