1,4-Dibromo-2-nitrobenzene ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,5-Dibromonitrobenzene
A10472.14 A10472.22
A10472.14EA 755 SEK
A10472.14 A10472.22
1,4-Dibromo-2-nitrobenzene ≥99%
1,4-Dibromo-2-nitrobenzene
Purity: 99% CAS No.: 3460-18-2 Molecular Formula: C6H3Br2NO2 Formula Weight: 280.91 Melting Point: 82-85° Beilstein Reference: 1950425
Formel: C₆H₃Br₂NO₂
MW: 280,9 g/mol
Smältpunkt: 82…85 °C
Densitet: 2,374
Storage Temperature: Omgivande
MDL Number: MFCD00007046
CAS nummer: 3460-18-2
EINECS: 222-404-8
UN: 3077
ADR: 9,III