4-Bromoaniline 96% pract.
Leverantör: Thermo Fisher Scientific
Fara
402871000 402875000
ACRO402871000EA 1050 SEK
ACRO402871000 ACRO402875000
4-Bromoaniline 96% pract.
4-Bromoaniline
Formel: C₆H₆BrN
Kokpunkt: 230…250 °C
Smältpunkt: 60…64 °C
Densitet: 1,497 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007822
CAS nummer: 106-40-1
EINECS: 203-393-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,01388
Order Now
Specifikation testresultat
Appearance Cream to grey to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 60 to 65 °C
GC ≥95 %