3-Bromoaniline ≥98%
Leverantör: Thermo Scientific
Fara
A13665.22 A13665.14 A13665.36
A13665.22EA 3575 SEK
A13665.22 A13665.14 A13665.36
3-Bromoaniline ≥98%
3-Bromoaniline
Purity: 98% CAS No.: 591-19-5 Molecular Formula: C6H6BrN Formula Weight: 172.03 Flash Point: 110°(230°F) Refractive Index: 1.6250 Odour: Characteristic Melting Point: 16° Boiling Point: 251° Beilstein Reference: 742028 UN No.: 2810 MDL No.: MFCD00007757
Formel: C₆H₆BrN
MW: 172,02 g/mol
Kokpunkt: 251 °C
Smältpunkt: 16 °C
Densitet: 1,590
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00007757
CAS nummer: 591-19-5
EINECS: 209-704-4
UN: 2810
ADR: 6.1,III