2,4-Dibromoaniline ≥98%
Leverantör: Thermo Fisher Scientific
Fara
A13329.06 A13329.14 A13329.22
A13329.06EA 630 SEK
A13329.06 A13329.14 A13329.22
2,4-Dibromoaniline ≥98%
2,4-Dibromoaniline
Purity: 98+% CAS No.: 615-57-6 Molecular Formula: C6H5Br2N Formula Weight: 250.93 Odour: Characteristic Melting Point: 78-82° Boiling Point: 156°/24mm Beilstein Reference: 2206653 UN No.: 2811 MDL No.: MFCD00007633
Formel: C₆H₅Br₂N
Kokpunkt: 156 °C (24 mmHg)
Smältpunkt: 78…82 °C
Densitet: 2,297
Storage Temperature: Omgivande
MDL Number: MFCD00007633
CAS nummer: 615-57-6
EINECS: 210-434-4
UN: 2811
ADR: 6.1,III