2,4,6-Tribromoaniline ≥98%
Leverantör: Thermo Scientific
Fara
A12453.22 A12453.36
A12453.22EA 1100 SEK
A12453.22 A12453.36
2,4,6-Tribromoaniline ≥98%
2,4,6-Tribromoaniline
Purity: 98% CAS No.: 147-82-0 Molecular Formula: C6H4Br3N Formula Weight: 329.83 Melting Point: 118-122° Boiling Point: 300° Merck Reference: 14,9610 MDL No.: MFCD00007634
Formel: C₆H₄Br₃N
MW: 329,82 g/mol
Kokpunkt: 300 °C
Smältpunkt: 118…122 °C
Densitet: 2,350
Storage Temperature: Omgivande
MDL Number: MFCD00007634
CAS nummer: 147-82-0
EINECS: 205-700-1
UN: 2811
ADR: 6.1,II
Merck Index: 14,9610