1-Bromo-3-iodobenzene 98+%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Bromoiodobenzene
249420100 249420500
ACRO249420100EA 1125 SEK
ACRO249420100 ACRO249420500
1-Bromo-3-iodobenzene 98+%
1-Bromo-3-iodobenzene
Formel: C₆H₄BrI
MW: 282,91 g/mol
Kokpunkt: 120 °C (18 mmHg)
Smältpunkt: –9,3 °C
Densitet: 2,21 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001043
CAS nummer: 591-18-4
EINECS: 209-703-9
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6590 to 1.6620 (20°C, 589 nm)
GC ≥98.0 %