4-Bromo-2-fluorophenol 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Bromo-3-fluoro-4-hydroxybenzene
293920250 293920050 293921000
ACRO293920250EA 1350 SEK
ACRO293920250 ACRO293920050 ACRO293921000
4-Bromo-2-fluorophenol 98%
4-Bromo-2-fluorophenol
Formel: C₆H₄BrFO
MW: 191 g/mol
Kokpunkt: 79 °C (7 mmHg)
Densitet: 1,74 g/cm³
Flampunkt: 98 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011722
CAS nummer: 2105-94-4
UN: 2810
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5660 to 1.5680 (20°C, 589 nm)
GC ≥97.5 %