1-Bromo-3-fluoro-4-iodobenzene 99+%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Fluoro-4-iodobromobenzene, 4-Bromo-2-fluoro-1-iodobenzene
249400250 249401000
ACRO249400250EA 1125 SEK
ACRO249400250 ACRO249401000
1-Bromo-3-fluoro-4-iodobenzene 99+%
1-Bromo-3-fluoro-4-iodobenzene
Formel: C₆H₃BrFI
Kokpunkt: 241 °C (1013 hPa)
Smältpunkt: 46…49 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010608
CAS nummer: 105931-73-5
Order Now
Specifikation testresultat
Appearance Off-white to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 46 to 49 °C
GC ≥99.0 %