4-Methoxybenzaldehyde dimethyl acetal ≥97.0%
Leverantör: TCI
Synonyms:
p-(Dimethoxymethyl)anisole, P-anisaldehyde dimethyl acetal, p-Anisaldehyde dimethyl acetal, 1-(Dimethoxymethyl)-4-methoxybenzene, 4-Methoxy-α,α-dimethoxytoluene
A1247-25ML
A1247-500ML
TCIAA1247-25MLEA
485
SEK
TCIAA1247-25ML
TCIAA1247-500ML
4-Methoxybenzaldehyde dimethyl acetal ≥97.0%
4-Methoxybenzaldehyde dimethyl acetal
Formel:
C₁₀H₁₄O₃ Kokpunkt: 253°C Flampunkt: 98°C |
MDL Number:
MFCD00036507 CAS nummer: 2186-92-7 EINECS: 218-577-4 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...