2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Leverantör: Merck
Synonyms:
2,4,6-Tri(2-pyridyl)-s-triazine, 2,4,6-Tri(2-pyridyl)-1,3,5-triazine, 2,4,6-Tris(2-pyridyl)-s-triazine
M-Clarity™ quality level = MQ100
T1253-1G
T1253-10G
T1253-25G
T1253-5G
T1253-1GEA
718
SEK
T1253-1G
T1253-10G
T1253-25G
T1253-5G
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
2,4,6-Tris(2-pyridyl)-1,3,5-triazine
Formel:
C₁₈H₁₂N₆ Storage Temperature: Omgivande |
MDL Number:
MFCD00006045 CAS nummer: 3682-35-7 EINECS: 222-965-9 |