1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate indicator for complexometry, Fluka™
Leverantör: Honeywell Chemicals
Varning
Synonyms: Catechol-3,5-disulphonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt monohydrate, Tiron monohydrate, 3,5-Pyrocatecholdisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulphonate monohydrate
89460-100G 89460-10G
89460-100GEA 2925 SEK
89460-100G 89460-10G
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate indicator for complexometry, Fluka™
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate
Formel: C₆H₄Na₂O₈S₂·1H₂O
MW: 332.22 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149531
CAS nummer: 270573-71-2
EINECS: 205-741-5
Order Now
Specifikation testresultat
Identity (IR) complying
Assay (HPLC) Min. 95.0 %
Indicator solution complying
Water (Karl Fischer) Max. 7 %
Suitability for Complexometry complying