3-Bromo-4-fluorobenzoic acid 96%
Leverantör: Thermo Scientific
Varning
B24711.03 B24711.06
B24711.03EA 1225 SEK
B24711.03 B24711.06
3-Bromo-4-fluorobenzoic acid 96%
3-Bromo-4-fluorobenzoic acid
Purity: 96% CAS No.: 1007-16-5 Molecular Formula: C7H4BrFO2 Formula Weight: 219.01 Melting Point: 138-141° MDL No.: MFCD00042463
Formel: C₇H₄BrFO₂
MW: 219,01 g/mol
Smältpunkt: 138…141 °C
Storage Temperature: Omgivande
MDL Number: MFCD00042463
CAS nummer: 1007-16-5
EINECS: 213-751-6