4-(4-Nitrobenzyl)pyridine, Sigma-Aldrich®
Leverantör: Merck
Synonyms: 4-(p-Nitrobenzyl)Pyridine
M-Clarity™ quality level = MQ200
8.20890.0025 8.20890.0100 8.20890.0005
8.20890.0025EA 2125 SEK
8.20890.0025 8.20890.0100 8.20890.0005
4-(4-Nitrobenzyl)pyridine, Sigma-Aldrich®
4-(4-Nitrobenzyl)pyridine
Formel: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00006445
CAS nummer: 1083-48-3
EINECS: 214-108-2
Order Now
Specifikation testresultat
Solubility (20 °C) insoluble
Melting Point 68 - 71 °C
Molar Mass 214.22 g/mol