Diisopropanolamin, Sigma-Aldrich®
Leverantör: Merck
Varning
M-Clarity™ quality level = MQ200
8.03264.0100 8.03264.1000 8.03264.9025
8.03264.0100EA 360 SEK
8.03264.0100 8.03264.1000 8.03264.9025
Diisopropanolamin, Sigma-Aldrich®
Diisopropanolamin
Formel: C₆H₁₅NO₂
Storage Temperature: Omgivande
MDL Number: MFCD00004531
CAS nummer: 110-97-4
EINECS: 203-820-9
Order Now
Specifikation testresultat
Solubility 870 g/l (20 °C)
Melting Point 38 - 44 °C
Molar Mass 133.19 g/mol
Boiling Point 249 °C (1013 hPa)
Vapor Pressure 0.02 hPa (20 °C)
Flash Point 135 °C
Refractive Index 1.4702 (16 °C, 589 nm)
Explosion Limit 1.6 - 8.0 %(V)
Density 0.99 g/cm3 (20 °C)
pH 11.4 (100 g/l, H2O, 20 °C)
Ignition Point 290 °C