2,4-Dichlorophenylhydrazinium chloride

Synonyms: (2,4-Dichlorophenyl)hydrazine hydrochloride, 2,4-Dichlorophenylhydrazine hydrochloride, 1-(2,4-Dichlorophenyl)hydrazine hydrochloride

BTB07732DA
MAYBBTB07732DAEA 0 SEK
MAYBBTB07732DA
2,4-Dichlorophenylhydrazinium chloride
2,4-Dichlorophenylhydrazinium chloride
Formel: C₆H₇Cl₃N₂
MW: 213,49 g/mol
MDL Number: MFCD00012929
CAS nummer: 5446-18-4
EINECS: 226-659-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR