Chlorodimethylvinylsilane 97%, rods
Leverantör: Thermo Fisher Scientific
|
Fara
|
Synonyms:
Dimethylvinylchlorosilane, Vinyldimethylchlorosilane
L16198.14
L16198.22
L16198.14EA
940
SEK
L16198.14
L16198.22
Chlorodimethylvinylsilane 97%, rods
Chlorodimethylvinylsilane
Purity: 97%
CAS No.: 1719-58-0
Molecular Formula: C4H9ClSi
Formula Weight: 120.66
Flash Point: -12°(10°F)
Refractive Index: 1.4160
Boiling Point: 82-83°
Beilstein Reference: 1737688
UN No.: 2985
MDL No.: MFCD00018090
Formel:
C₄H₉ClSi MW: 120.65 g/mol Kokpunkt: 82…83 °C Densitet: 0.883 Flampunkt: –12 °C (10 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00018090 CAS nummer: 1719-58-0 EINECS: 217-007-1 UN: 2985 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...