1,2-Dibenzoylethane ≥98%
Leverantör: Thermo Scientific
Synonyms: 1,4-Diphenyl-1,4-butanedione, 1,4-Diphenylbutane-1,4-dione
L08652.03 L08652.06
L08652.03EA 1400 SEK
L08652.03 L08652.06
1,2-Dibenzoylethane ≥98%
1,2-Dibenzoylethane
Purity: 98+% CAS No.: 495-71-6 Molecular Formula: C16H14O2 Formula Weight: 238.29 Melting Point: 144-148° Boiling Point: 260°/15mm MDL No.: MFCD00037818
Formel: C₁₆H₁₄O₂
MW: 238,29 g/mol
Kokpunkt: 260 °C (15 mmHg)
Smältpunkt: 144…148 °C
Storage Temperature: Omgivande
MDL Number: MFCD00037818
CAS nummer: 495-71-6