Ethylammonium chloride 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Aminoethane hydrochloride, Ethylamine hydrochloride
L03624.30 L03624.0B
L03624.30EA 1075 SEK
L03624.30 L03624.0B
Ethylammonium chloride 98%
Ethylammonium chloride
Observera: Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Formel: C₂H₈ClN
MW: 81,54 g/mol
Smältpunkt: 95…100 °C
Densitet: 1,220
Storage Temperature: Omgivande
MDL Number: MFCD00012885
CAS nummer: 557-66-4
EINECS: 209-182-8
Merck Index: 14,3762